Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC- Coated Amber Glass Bottle
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Formula Weight 58.08
Grade Meets ACS reagent chemical requirements
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point -20°C (-4°C)
Quantity 500mL
Chemical Name or Material Acetone