Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC- Coated Amber Glass Bottle

Chemical Identifiers

67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
CHEBI:15347

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
SMILES CC(=O)C
IUPAC Name propan-2-one
Formula Weight 58.08
Packaging Narrow Mouth PVC- Coated Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point −20.00°C
Boiling Point ≤56.1°C
Quantity 4L
Chemical Name or Material Acetone
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements