Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone Benzothiazolyl-2-hydrazone 98.0+%, TCI America™

Click to view available options
Quantity:
25g
6277-26-5
C10H11N3S
205.279
MFCD00059701
TZHQLYDPJQZYNF-UHFFFAOYSA-N
N-(2-Benzothiazolyl)acetonehydrazone
234769
N-(propan-2-ylideneamino)-1,3-benzothiazol-2-amine
CC(=NNC1=NC2=CC=CC=C2S1)C
6277-26-5
C10H11N3S
205.279
MFCD00059701
TZHQLYDPJQZYNF-UHFFFAOYSA-N
N-(2-Benzothiazolyl)acetonehydrazone
234769
N-(propan-2-ylideneamino)-1,3-benzothiazol-2-amine
CC(=NNC1=NC2=CC=CC=C2S1)C
Quantity 25g
Color White-Yellow
Melting Point 198°C
Formula Weight 205.28
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Acetone Benzothiazolyl-2-hydrazone