Alfa Aesar™ Acetone-d6, 100%(Isotopic)

Manufacturer: Alfa Aesar™ 42262AA

Quantity 5mL

 View more versions of this product

Catalog No. AA42262AA

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 666-52-4
    Formula Weight 64.13
    MDL Number MFCD00044635
    UN Number UN1090
    Chemical Name or Material Acetone-d6, Isotopic

    Specifications

    Formula Weight 64.13
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point −94°C
    Physical Form Liquid
    Quantity 5mL
    Odor Acetone-like
    Density 0.872g/mL
    Boiling Point 56°C
    Color Colorless
    Flash Point −17° (1°F)
    Refractive Index 1.3554
    MDL Number MFCD00044635
    CAS 666-52-4
    UN Number UN1090
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONE
    Chemical Name or Material Acetone-d6, Isotopic
    Assay 100%

    GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit