Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone - D6, deuteration degree min. 99.9% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Click to view available options
Quantity:
10mL
25mL
100mL
10 x 0.5mL
10 x 0.75mL
Packaging:
Glass ampoule
Glass bottle
Septum ampoule
666-52-4
C3H6O
64.117
CSCPPACGZOOCGX-WFGJKAKNSA-N
acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone
522220
CHEBI:78217
1,1,1,3,3,3-hexadeuteriopropan-2-one
CC(=O)C
666-52-4
C3H6O
64.117
CSCPPACGZOOCGX-WFGJKAKNSA-N
acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone
522220
CHEBI:78217
1,1,1,3,3,3-hexadeuteriopropan-2-one
CC(=O)C

For NMR Spectroscopy

Viscosity 0.32mPa·s (20°C/68°F)
Ignition Point 465°C (869°F)
Quantity 10 x 0.5mL
Odor Characteristic
Packaging Glass ampoule
Vapor Pressure 233 hPa at 20°C (68°F)
Color Colorless
Assay Percent Range 99.9% min.
Formula Weight 64.12
Physical Form Liquid
Chemical Name or Material Acetone - D6
Show More Show Less