Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone-d6, for NMR, +99% atom D, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
666-52-4
C3H6O
64.117
MFCD00044635
CSCPPACGZOOCGX-WFGJKAKNSA-N
acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone
522220
CHEBI:78217
1,1,1,3,3,3-hexadeuteriopropan-2-one
CC(=O)C
666-52-4
C3H6O
64.117
MFCD00044635
CSCPPACGZOOCGX-WFGJKAKNSA-N
acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone
522220
CHEBI:78217
1,1,1,3,3,3-hexadeuteriopropan-2-one
CC(=O)C
Color Colorless
Packaging Glass bottle
Quantity 10mL
Deuterium (D) 99.0% min.
Density 0.8700g/mL
Boiling Point 55.0°C
Melting Point -93.0°C
Linear Formula CD3COCD3
Flash Point −20°C
Beilstein 01, III, 2741
Formula Weight 64.13
Physical Form Liquid
Grade NMR
Chemical Name or Material Acetone-d6, 99+ atom % D
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-563-9