Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Electronic/Cleanroom Grade, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18608385 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 gal.
Packaging:
Poly Bottle

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Alkalinity 0.5 μeq/g
Acidity 0.3 μeq/g
CAS Max % 1
Boiling Point 56°C
Packaging Poly Bottle
Quantity 1 gal.
Residue after Evaporation 5 ppm
Percent Purity 0.995
Grade Electronic/Cleanroom Grade
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.