Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone, Exceeds A.C.S. Specifications, HPLC Grade, 99.5%, Spectrum™ Chemical
Click to view available options
Quantity:
4 L
4 x 4 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Acetone, Exceeds A.C.S. Specifications, HPLC Grade solvents are lot-tested and precisely controlled to meet the high quality requirements of chromatographic analysis including high purity, low UV-absorbance, guaranteed suitability for fluorescence detection, low particulates (eliminates pre-filtration) and controlled low-content of non-volatile components. All Acetone chromatography solvents are delivered with an expiration date.
Specifications
CAS Max % | 100% |
Quantity | 4 L |
Packaging | Amber Glass Bottle |
Residue after Evaporation | 5 ppm |
Titratable Acid | 0.3 μeq/g |
Titratable Base | 0.6 μeq/g |
UN Number | UN1090 |
Water | 0.5% |
Percent Purity | 99.5% |
Grade | High Purity |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction