Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Exceeds A.C.S. Specifications, HPLC Grade, 99.5%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O

HP402, 67-64-1, C3H6O

Spectrum™ Acetone, Exceeds A.C.S. Specifications, HPLC Grade solvents are lot-tested and precisely controlled to meet the high quality requirements of chromatographic analysis including high purity, low UV-absorbance, guaranteed suitability for fluorescence detection, low particulates (eliminates pre-filtration) and controlled low-content of non-volatile components. All Acetone chromatography solvents are delivered with an expiration date.
CAS Max % 100%
Aldehyde 0.002%
UN Number UN1090
Percent Purity 99.5%
Grade High Purity Solvent
Methanol 0.05%
Packaging Amber Glass Bottle
Residue after Evaporation 5 ppm
Titratable Acid 0.3 μeq/g
Titratable Base 0.6 μeq/g
Water 0.5%
Quantity 1 L
Show More Show Less