Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone, FCC, 99.5-100.5%, Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
Description
Spectrum™ Chemical Acetone, FCC also known as 2-propanone, is an organic compound and a colorless, flammable liquid. Due to its miscibility with water, it serves as an important solvent. The U.S. Food and Drug Administration (FDA) has determined acetone is safe for use as an indirect food additive in adhesives and food-contact coatings and is regarded as a Generally Recognized as Safe (GRAS) substance at certain concentrations. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Acidity | 0.002% |
Alkalinity | 10 mg/kg |
Quantity | 4 L |
Packaging | Poly Bottle |
Refractive Index | 1.358-1.360 |
UN Number | UN1090 |
Water | 0.5% |
Percent Purity | 99.5 to 100.5% |
Grade | FCC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction