Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, FCC, 99.5-100.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O

A1041, 67-64-1, C3H6O

Spectrum™ Acetone, FCC also known as 2-propanone, is an organic compound and a colorless, flammable liquid. Due to its miscibility with water, it serves as an important solvent. The U.S. Food and Drug Administration (FDA) has determined acetone is safe for use as an indirect food additive in adhesives and food-contact coatings and is regarded as a Generally Recognized as Safe (GRAS) substance at certain concentrations. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Acidity 0.002%
Alkalinity 10 mg/kg
Aldehyde 0.002%
UN Number UN1090
Percent Purity 99.5 to 100.5%
Lead (Pb) 1 mg/kg
Methanol 0.05%
Non_volatile Matter 10 mg/kg
Packaging Amber Glass Bottle
Refractive Index 1.358-1.360
Specific Gravity at 25 degC 0.788
Water 0.5%
Quantity 4 x 4 L
Show More Show Less