Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Used in histology applications

Packaging Poly Bottle
Quantity 4L

 View more versions of this product

Catalog No. A16P-4



 

Due to product restrictions, please to purchase or view availability for this product.

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

67-64-1
Assay: ≥99.5%
CH3COCH3
MFCD00008765
Acetone
Liquid
C3H6O
58.08
2-Propanone
Histological Grade

Specifications

67-64-1
Liquid
C3H6O
58.08
2-Propanone
Poly Bottle
0.32 mPa/s at 20°C
56°C
−95°C
4L
Histological Grade
Vapor Pressure: 247mbar at 20°C
Assay: ≥99.5%
CH3COCH3
MFCD00008765
2.0 (Air = 1.0)
247mbar at 20°C
>4°C
Colorless
7
Acetone

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit