Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone (Histological), Fisher Chemical™
Click to view available options
Quantity:
1 gal.
4 L
20 L
Packaging:
Aluminum Can
Poly Bottle
Steel Pail
Specifications
pH | 7 |
Melting Point | -95°C |
Color | Colorless |
Decomposition Information | >4°C |
Density | 0.790g/cm³ |
Boiling Point | 56°C |
Quantity | 4 L |
Packaging | Aluminum Can |
Physical Form | Liquid |
Assay Percent Range | Assay: ≥99.5% |
Show More |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. NFPA Health:1 Flammability:3 Instability:0 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction