Acetone (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

HPLC mobile phase for analytical and preparative separations

Packaging NOWPAK I (Plastic Exterior)
Quantity 19L

 View more versions of this product

Catalog No. A949N-119

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 67-64-1
    Assay Percent Range 99.5% min by GC
    Molecular Formula C3H6O
    Linear Formula CH3COCH3
    Formula Weight 58.08
    MDL Number MFCD00008765
    Synonym 2-Propanone
    Chemical Name or Material Acetone
    Assay ≥99.5%
    Grade HPLC Grade, Mobile phase for HPLC applications

    Specifications

    Boiling Point 56°C
    Melting Point −95°C
    Formula Weight 58.08
    Quantity 19L
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open f
    DOT Information DOT Class 3, : Flammable Liquid
    Physical Form Liquid
    Color Colorless
    pH 7
    Density 0.790g/cm³
    Viscosity 0.32 mPa/s at 20°C
    Decomposition Information >4°C
    Vapor Pressure 247mbar at 20°C
    Isopropyl Alcohol 0.05% max.
    Additional Information Vapor Pressure: 247mbar at 20°C
    Methanol 0.05% max.
    Solubility in Water Pass Test
    Aldehyde 0.002% max.
    Assay Percent Range 99.5% min by GC
    Absorbance 0.01 AU max. at 400nm
    Synonym 2-Propanone
    Refractive Index 1.3555 to 1.3580 (at 25°C)
    Cleanup Symbol Solvent
    MDL Number MFCD00008765
    Health Hazard 2 DANGER!
    Identification Pass Test
    Packaging NOWPAK I (Plastic Exterior)
    Residue after Evaporation 1ppm max.
    Substances Reducing Permanganate Pass Test
    Titratable Acid 0.0003mEq/g max.
    Titratable Base 0.0006mEq/g max.
    Water 0.5% max.
    Molecular Formula C3H6O
    Linear Formula CH3COCH3
    CAS 67-64-1
    ChemAlert Storage Symbol Red
    Fluorescence Background Pass Test
    Chemical Name or Material Acetone
    Assay ≥99.5%
    Grade HPLC Grade, Mobile phase for HPLC applications
       

    DANGER!

    Emergency Overview
    Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open f

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit