Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, HPLC Grade, 99.5+%, Alfa Aesar™

Click to view available options
Quantity:
1L
4L
4 x 1L
4 x 4L
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O
Assay Percent Range >99.5%
Appearance Colorless liquid
Beilstein 635680
Solubility Information Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils.
Formula Weight 58.08
Physical Form Liquid
Grade HPLC
Density 0.791
Odor Characteristic
Refractive Index 1.359
Vapor Pressure 247 hPa (185mm Hg) at 20°C
Flash Point -17°C (1°F)
Boiling Point 56°C
Color Colorless
Melting Point °C to 94°C
Quantity 1L
Chemical Name or Material Acetone
Show More Show Less

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H336-H373

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-662-2

RTECSNumber : AL3150000

TSCA : Yes

Recommended Storage : Ambient temperatures