Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, HPLC Grade, 99.5%, Spectrum™

Click to view available options
Quantity:
200 L
Packaging:
Drum
CAS 67-64-1
CAS Max % 1
Aldehyde 2e-05
Percent Purity 0.995
Grade HPLC
Isopropyl Alcohol 0.0005
Packaging Drum
Residue after Evaporation 1e-05
Solubility in Water To Pass Test
Titratable Acid 0.3 μeq/g
Titratable Base 0.6 μeq/g
Water 0.005
Quantity 200 L
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.