Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, HPLC Grade, Safe-Cote, Fisher Chemical™

Click to view available options
Quantity:
4 L
CAS 67-64-1
Molecular Formula C3H6O
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Grade Chromatography
Packaging Safe-Cote™ Amber Glass 
Quantity 4 L
Show More Show Less