Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Meets or Exceeds A.C.S. Specifications, SpectroSolv (TM), 99.5%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
4 x 4 L
Packaging:
Amber Glass Bottle
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O

SP403, 67-64-1, C3H6O

Spectrum™ Acetone, Meets or Exceeds A.C.S. Specifications, SpectroSolv (TM) also known as propanone, is an organic compound and a colorless, flammable liquid. Due to its miscibility with water, it serves as an important solvent. Used as a polar aprotic solvent in a wide range of organic reactions, acetone does not form an azeotrope with water. Thanks to it′s low cost and volatility, it is also commonly used to clean laboratory glassware. Another characteristic of acetone is that it can be cooled with dry ice to -78 C without freezing. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.
CAS Max % 100%
Aldehyde 0.002%
UN Number UN1090
Percent Purity 99.5%
Methanol 0.05%
Packaging Amber Glass Bottle
Residue after Evaporation 10 ppm
Titratable Acid 0.3 μeq/g
Titratable Base 0.6 μeq/g
Water 0.5%
Quantity 6 x 500 mL
Show More Show Less