Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Multi-Compendial, N.F., J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
CHEBI:15347
SMILES CC(=O)C
IUPAC Name propan-2-one
Formula Weight 58.08
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point −20.00°C
Quantity 500mL
Chemical Name or Material Acetone
Grade Multi-Compendial/NF