Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, NF, 99%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O

AC115, 67-64-1, C3H6O

Spectrum™ Acetone, NF is a low boiling, volatile organic compound. It is considered one of the five common excipient solvents suitable for pharmaceutical preparations, including water, methanol, dehydrated alcohol and ether. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN1090
Percent Purity 99%
Non_volatile Matter 0.004%
Packaging Amber Glass Bottle
Specific Gravity at 25 degC 0.789
Water 0.5%
Quantity 500 mL