Acetone (NF/FCC/EP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Steel Pail
Quantity 20L

 View more versions of this product

Catalog No. A9-20

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 67-64-1
    Molecular Formula C3H6O
    Linear Formula CH3COCH3
    Formula Weight 58.08
    MDL Number MFCD00008765
    Synonym 2-Propanone
    Chemical Name or Material Acetone
    Assay NLT 99.5%, NMT 100.5%% (FCC)
    Grade BP/EP/FCC/JP/NF/USP

    Specifications

    Boiling Point 56°C
    Melting Point −95°C
    Formula Weight 58.08
    Quantity 20L
    UN Number 1090; Colorless
    DOT Information DOT Class 3, : Flammable Liquid
    Physical Form Liquid
    Color Colorless
    pH 7
    Viscosity 0.32 mPa/s at 20°C
    Decomposition Information >4°C
    Specific Gravity 0.7880 max. (FCC)
    Density 2.0 (Air = 1.0)
    Vapor Pressure 247mbar at 20°C
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

    NFPA

    Health:1
    Flammability:3
    Instability:0
    Non_volatile Matter 0.004% max. (NF)
    Readily Oxidizable Substances Pass Test (NF)
    Water ≤3g/L (EP)
    Identification Pass Test
    Isopropyl Alcohol ≤0.05% (EP)
    Lead (Pb) 1mg/kg max. (FCC)
    Methanol 0.05% max. (EP)
    Packaging Steel Pail
    Phenols Pass Test (FCC)
    Refractive Index 1.358 to 1.360 (at 25°C)
    Residual Solvents Meets Requirements (BP)
    Alkalinity 10mg/kg max. (FCC)
    Residue after Evaporation 50ppm max. (EP)
    Aldehyde 0.002% max. (FCC)
    Substances Reducing Permanganate Pass Test (FCC)
    Benzene 2ppm max. (EP)
    Synonym 2-Propanone
    Distillation Range 1.0°C max. (FCC)
    Acidity Alkalinity Pass Test (EP)
    Reducing Substances Pass Test (EP)
    MDL Number MFCD00008765
    ChemAlert Storage Symbol Red
    Health Hazard 2 DANGER!
    Distillation Temperature 55.1° to 57.1°C (FCC)
    Insoluble in Water Pass Test (EP)
    Additional Information Vapor Pressure: 247mbar at 20°C
    Solubility in Water Pass Test (FCC)
    Cleanup Symbol Solvent
    Any Other Impurity 0.05% max. (EP)
    Appearance of Solution Pass Test (EP)
    Molecular Formula C3H6O
    Linear Formula CH3COCH3
    CAS 67-64-1
    Acetic Acid 0.002% max. (FCC)
    Chemical Name or Material Acetone
    Assay NLT 99.5%, NMT 100.5%% (FCC)
    Grade BP/EP/FCC/JP/NF/USP
       

    DANGER!

    Emergency Overview
    Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

    NFPA

    Health:1
    Flammability:3
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit