Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Packaged under Nitrogen, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle

Chemical Identifiers

67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
CHEBI:15347
propan-2-one
CC(=O)C

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory, research, or manufacturing use
Formula Weight 58.08
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point −20.00°C
Quantity 4L
Chemical Name or Material Acetone, Packaged under Nitrogen
Grade HPLC Grade