Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetone, Reagent, ACS, 99.5%, Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
Description
Spectrum™ Chemical Acetone, Reagent, ACS is colorless, flammable, liquid. Due to it′s micsibilaty with water, it serves as an important solvent. Used as a polar aprotic solvent in a wide range of organic reactions, acetone does not form an azeotrope with water. Thanks to it′s low cost and volatility, it is also commonly used to clean laboratory glassware. Another characteristic of Acetone is that it can be cooled with dry ice to -78°C without freezing allowing this solvent to be used to conduct reactions at low temperature. As an ACS grade Reagent, Spectrum Chemical manufactured Acetone is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Specifications
CAS Max % | 100% |
Quantity | 500 mL |
Packaging | Amber Glass Bottle |
Residue after Evaporation | 0.001% |
Titratable Acid | 0.0003 meq/g |
Titratable Base | 0.0006 meq/g |
UN Number | UN1090 |
Water | 0.5% |
Percent Purity | 99.5% |
Grade | ACS |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction