Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Acetone semicarbazone, 96%

Manufacturer:  Alfa Aesar™ A1243622

Quantity 100g

 View more versions of this product

Catalog No. AAA1243622


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

110-20-3
115.14
970110
Acetone semicarbazone
C4H9N3O
MFCD00014785
14,8444

Specifications

110-20-3
115.14
970110
179° to 185°C
Acetone semicarbazone
C4H9N3O
MFCD00014785
14,8444
100g
96%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : AL7175000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit