Alfa Aesar™ Acetone semicarbazone, 96%

Manufacturer: Alfa Aesar™ A1243618

Quantity 50g

 View more versions of this product

Catalog No. AAA1243618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-20-3
    Molecular Formula C4H9N3O
    Formula Weight 115.14
    MDL Number MFCD00014785
    Beilstein 970110
    Merck Index 14,8444
    Chemical Name or Material Acetone semicarbazone

    Specifications

    Formula Weight 115.14
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C4H9N3O
    Beilstein 970110
    RTECS Number AL7175000
    MDL Number MFCD00014785
    Merck Index 14,8444
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    TSCA TSCA
    CAS 110-20-3
    Melting Point 179° to 185°C
    Quantity 50g
    Chemical Name or Material Acetone semicarbazone
    Assay 96%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit