Acetone, Semiconductor Grade, 99.5%, Alfa Aesar

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Acidity 0.3µeq/g, Alkalinity 0.5µeq/g, H2O 0.5%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.1ppm

Manufacturer: Alfa Aesar 19392K2

Quantity 1L

 View more versions of this product

Catalog No. AA19392K2

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 67-64-1
    Physical Form Liquid
    Assay Percent Range 99.5%
    Molecular Formula C3H6O
    Formula Weight 58.08
    MDL Number MFCD00008765
    Chemical Name or Material Acetone
    Grade Semiconductor Grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Formula Weight 58.08
    Quantity 1L
    DOT Information Hazard Class: 3, Packing Group: II
    EINECS Number 200-662-2
    Ignition Point 465°C (869°F)
    Odor Characteristic
    Vapor Pressure 247 hPa (185mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range 99.5%
    Molecular Formula C3H6O
    MDL Number MFCD00008765
    CAS 67-64-1
    Health Hazard 1 H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.
    Chemical Name or Material Acetone
    Grade Semiconductor Grade

    H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit