Acetone, Spectrophotometric Grade, 99.5+%, Alfa Aesar

Manufacturer: Alfa Aesar 32451K7

Quantity 4L

 View more versions of this product

Catalog No. AA32451K7

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 67-64-1
    Physical Form Liquid
    Assay Percent Range >99.5%
    Molecular Formula C3H6O
    Formula Weight 58.08
    MDL Number MFCD00008765
    Chemical Name or Material Acetone
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 58.08
    Quantity 4L
    Color Colorless
    DOT Information DOT Class: 3; Packing Group: II
    EINECS Number 200-662-2
    Physical Form Liquid
    Appearance Colorless liquid
    MDL Number MFCD00008765
    Odor Characteristic
    Vapor Pressure 247 hPa (185mm Hg) at 20°C
    Assay Percent Range >99.5%
    Molecular Formula C3H6O
    CAS 67-64-1
    Health Hazard 1 H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.
    Chemical Name or Material Acetone
    Grade Spectrophotometric grade

    H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit