Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Technical, Fisher Chemical

Click to view available options
Quantity:
20L
200L
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C
67-64-1
C3H6O
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone
180
CHEBI:15347
propan-2-one
CC(=O)C
Fisher Chemical recommends that this product is used where purity requirements are less stringent, such as glassware and laboratory clean up.
Appearance Clear, colorless liquid
UN Number 1090
Formula Weight 58.08
Physical Form Liquid
Percent Purity ≤98%
Grade Technical
Density 0.790
Identification Pass Test
Odor Sweet
Packaging Steel pail
Refractive Index 1.358 to 1.359
Solubility in Water Soluble
Vapor Pressure 247mbar at 20°C
Vapor Density 2.0
Flash Point -20°C (-4°F); Closed cup
Viscosity 0.32mPa.s at 20°C
Boiling Point 56°C
Color Undesignated
Melting Point -95°C
pH 7
Quantity 20L
Chemical Name or Material Acetone
Show More Show Less

Recommended Storage :

  • Flammables area
  • Keep containers tightly closed in a dry, cool and well-ventilated place
  • Keep away from heat and sources of ignition
  • Keep container tightly closed in a dry and well-ventilated place