Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, UniSolv(R), 99.9%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
67-64-1
67-64-1
CAS Max % 1
Aldehyde 2e-05
Percent Purity 0.999
Grade High Purity Solvent
Fluorescence Background 1 ppb
Isopropyl Alcohol 0.0005
Methanol 0.0005
Packaging Amber Glass Bottle
Residue after Evaporation 1 ppm
Solubility in Water To Pass Test
Titratable Acid 0.0003 meq/g
Titratable Base 0.0006 meq/g
Water 0.005
Quantity 1 L
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.