Acetone, World/GMP, ACS/USP/NF/BP/EP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 329WORLDCS4L

Packaging Amber Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. 16-101-413

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 67-64-1
    Physical Form Liquid
    Molecular Formula C3H6O
    Formula Weight 58.08
    Synonym 2-Propanone
    Chemical Name or Material Acetone

    Specifications

    Formula Weight 58.08
    Quantity 4L
    Color Colorless, invisible vapor
    Packaging Amber Glass Bottle
    Flash Point -20°C (-4°C)
    Physical Form Liquid
    Odor Sweet, alcohol-like
    Stability Stable under recommended storage conditions.
    Molecular Formula C3H6O
    Synonym 2-Propanone
    CAS 67-64-1
    Description General purpose organic solvent
    Chemical Name or Material Acetone

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit