Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, 0.1% Formic Acid; Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
CHEBI:38472

Function tested and optimized for LC/MS applications

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Linear Formula CH3CN
UN Number 1648; Colorless
SMILES CC#N
IUPAC Name acetonitrile
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point 2°C (35.8°F)
Boiling Point 81°C to 82°C
Melting Point -46°C
Quantity 4L
Chemical Name or Material Acetonitrile, 0.1% Formic Acid; Filtered through a 0.2 micron filter
Grade BAKER ANALYZED™ LC-MS Reagent
Show More Show Less