Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, 0.1% Formic Acid; Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T. Baker™

Catalog No. p-5383224
Click to view available options
Quantity:
1 L
4 L
Packaging:
Narrow Mouth Amber Glass Bottle

Function tested and optimized for LC/MS applications

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Quantity 1 L
Flash Point 2°C (35.8°F)
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Linear Formula CH3CN
UN Number 1648; Colorless
Grade BAKER ANALYZED™ LC-MS Reagent
Chemical Name or Material Acetonitrile, 0.1% Formic Acid; Filtered through a 0.2 micron filter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.