Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile with 0.1% (v/v) Acetic Acid, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™

Catalog Number 01100255
Click to view available options
Quantity:
2.5 L
4 L
Melting Point -45.7°C
Grade Hypergrade
Boiling Point 81.6°C (1013 hPa)
Quantity 4 L
Explosion Limit 3 to 17% (V) (refers to pure substance)
Flash Point 2 °C (refers to pure substance)
Ignition Point 524 °C (refers to pure substance)
Packaging Glass Bottle
Vapor Pressure 97 hPa at 20 °C
Recommended Storage Store at 2 to 30 °C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit