Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetonitrile with 0.1% (v/v) Formic Acid, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™
Click to view available options
Quantity:
1 L
2.5 L
4 L
Specifications
Melting Point | -45.7°C |
Grade | Hypergrade |
Density | 0.786 g/cm3 at 20 °C |
Boiling Point | 81.6°C (1013 hPa) |
Quantity | 2.5 L |
Explosion Limit | 3 to 17% (V) |
Flash Point | 2 °C |
Ignition Point | 524 °C |
Packaging | Glass Bottle |
Vapor Pressure | 97 hPa at 20 °C |
Safety and Handling
Recommended Storage | Store at 2 to 30 °C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction