Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetonitrile 90% and Water 10% with 1% formic acid, LabReady™ Solvent Blend, for HPLC, Burdick & Jackson, Honeywell
Click to view available options
Quantity:
4 x 4 L
Description
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
Specifications
CAS Max % | 86.30, 12.20, 1.50 |
Grade | HPLC |
Density | 0.8069 g/cm3 |
Boiling Point | 66.1°C |
Quantity | 4 x 4 L |
Flash Point | 8.3°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Safety and Handling
H225+H302+H332+H315+H319+H370
P280+P271+P270+P264+P260+P243+P242+P241+P240+P233+P210
Danger
Recommended Storage : Room Temperature