Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
Additional Information Vapor Pressure: 97mbar at 20°C
Assay Percent Range 99.9%
Linear Formula CH3CN
UN Number 1648; Colorless
Beilstein 02, 183
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Formula Weight 41.04
Percent Purity ≥99.9%
Grade Extra Dry
Density 0.7810g/mL
Identification Pass Test
Infrared Spectrum Conforms to reference
Packaging AcroSeal™
Residue after Evaporation 1ppm max.
Water 50ppm max.
Flash Point 2°C (35.8°F)
Decomposition Information >500°C
Melting Point -46°C
Quantity 100mL
Chemical Name or Material Acetonitrile, AcroSeal™, Quadrant Cap
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger