Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
Packaging:
Glass Bottle
Plastic Jerrycan
Plastic drum
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
Additional Information Vapor Pressure: 97mbar at 20°C
Assay Percent Range 99+%
Linear Formula CH3CN
Beilstein 02, 183
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Formula Weight 41.05
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Density 0.7810g/mL
Packaging Glass Bottle
Refractive Index 1.3430 to 1.3450
Residue after Evaporation 0.005% max.
Solubility Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Specific Gravity 0.781
Water 0.3% max. (K.F.)
Flash Point 2°C
Viscosity 0.36 mPa.s (20°C)
Decomposition Information >500°C
Boiling Point 81.0°C to 82.0°C
Color Undesignated
Melting Point -46.0°C
Quantity 2.5L
Chemical Name or Material Acetonitrile
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger