Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, anhydrous, >99.8%, Alfa Aesar™

Click to view available options
Quantity:
250mL
1L
4L
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N

Water <100ppm, typically 50ppm (0.005%)

Assay Percent Range >99.8%
Appearance Colorless liquid
UN Number 1648; Colorless
Beilstein 741857
Solubility Information Miscible with water,alcohols,ethers,acetone,chloroform,carbon tetrachloride and ethylene chloride. Immiscible with saturated hydrocarbons (petroleum fractions).
Formula Weight 41.05
Physical Form Liquid
Density 0.786 g/cm3 at 20°C
Ignition Point 525°C (977°F)
Refractive Index 1.344
Vapor Pressure 97 hPa (73mm Hg) at 20°C
Flash Point 5°C (41°F)
Boiling Point 80°C to 82°C
Color Colorless
Melting Point °C to 48 to°C to 45°C
Quantity 250mL
Chemical Name or Material Acetonitrile
Show More Show Less

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c

H225-H302+H312+H332-H319

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 200-835-2

RTECSNumber : AL7700000

TSCA : Yes

Recommended Storage : Ambient temperatures