Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, BAKER ANALYZED™ HPLC ULTRA Gradient Solvent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
Formula Weight 41.05
Grade HPLC
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Flash Point 2°C (35.8°F)
Quantity 4L
Chemical Name or Material Acetonitrile