Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, BAKER ANALYZED™ ULTRA LC/MS, J.T.Baker™

Click to view available options
Quantity:
2 x 1L
6 x 1L
Packaging:
Narrow Mouth Flint
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N

For cutting-edge LC/MS applications

    For use in LC/MS and UHPLC
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Meets ACS reagent chemical requirements
Packaging Narrow Mouth Flint
Specific Gravity 1L = 0.79kg
Flash Point 2.00°C
Quantity 2 x 1L
Chemical Name or Material Acetonitrile