Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Acetonitrile-d3, 99.8% (Isotopic)

Manufacturer:  Alfa Aesar™ 4226404

Quantity 2g

 View more versions of this product

Catalog No. AA4226404


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2206-26-0
MFCD00001881
14,70
44.08
UN1648
Acetonitrile-d3, Isotopic

Specifications

2206-26-0
Hygroscopic
81°C
2g
5° (42°F)
MFCD00001881
14,70
99.8%
Pungent
0.844g/mL
Liquid
Colorless
44.08
UN1648
Acetonitrile-d3, Isotopic

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H302-H312-H332-H319
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit