Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile - D3, deuteration degree min. 99.8% for NMR spectroscopy, MagniSolv, MilliporeSigma™

Click to view available options
Quantity:
10mL
Packaging:
Septum ampoule
2206-26-0
C2H3N
44.071
WEVYAHXRMPXWCK-FIBGUPNXSA-N
acetonitrile-d3,2h3 acetonitrile,cd3cn,acetonitrile-2,2,2-d3,methyl-d3 cyanide,trideuteroacetonitrile,acetonitrile-d3, 99.8 atom % d,acetonitrile-d3, >=99.8 atom % d,acetonitrile-d isotopic 5g
123151
2,2,2-trideuterioacetonitrile
CC#N
2206-26-0
C2H3N
44.071
WEVYAHXRMPXWCK-FIBGUPNXSA-N
acetonitrile-d3,2h3 acetonitrile,cd3cn,acetonitrile-2,2,2-d3,methyl-d3 cyanide,trideuteroacetonitrile,acetonitrile-d3, 99.8 atom % d,acetonitrile-d3, >=99.8 atom % d,acetonitrile-d isotopic 5g
123151
2,2,2-trideuterioacetonitrile
CC#N

For NMR Spectroscopy

Viscosity 0.39mPa·s (20°C)
Packaging Septum ampoule
Ignition Point 525°C (977°F)
Quantity 10mL
Odor Ether - like
Vapor Pressure 97 hPa at 20°C (68°F)
Assay Percent Range 99.8% min.
Formula Weight 44.07
Physical Form Liquid
Chemical Name or Material Acetonitrile - D3