Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile-d3, for NMR, 99% atom D, ACROS Organics™

Click to view available options
Quantity:
10mL
25mL
50mL
250mL
Packaging:
Glass bottle
2206-26-0
C2H3N
44.071
MFCD00001881
WEVYAHXRMPXWCK-FIBGUPNXSA-N
acetonitrile-d3,2h3 acetonitrile,cd3cn,acetonitrile-2,2,2-d3,methyl-d3 cyanide,trideuteroacetonitrile,acetonitrile-d3, 99.8 atom % d,acetonitrile-d3, >=99.8 atom % d,acetonitrile-d isotopic 5g
123151
2,2,2-trideuterioacetonitrile
CC#N
2206-26-0
C2H3N
44.071
MFCD00001881
WEVYAHXRMPXWCK-FIBGUPNXSA-N
acetonitrile-d3,2h3 acetonitrile,cd3cn,acetonitrile-2,2,2-d3,methyl-d3 cyanide,trideuteroacetonitrile,acetonitrile-d3, 99.8 atom % d,acetonitrile-d3, >=99.8 atom % d,acetonitrile-d isotopic 5g
123151
2,2,2-trideuterioacetonitrile
CC#N

Acetonitrile-d3, C2H3N, CAS Number-2206-26-0, 2h3 acetonitrile, acetonitrile-2,2,2-d3, acetonitrile-d3, acetonitrile-d3, >-99.8 atom % d, acetonitrile-d3, 99.8 atom % d, acetonitrile-d isotopic 5g, cd3cn, methyl-d3 cyanide, trideuteroacetonitrile, 10mL, 02, IV, 428, 79.0 deg.C to 81.0 deg.C

Acetonitrile-d3, C2H3N, CAS Number-2206-26-0, 2h3 acetonitrile, acetonitrile-2,2,2-d3, acetonitrile-d3, acetonitrile-d3, >-99.8 atom % d, acetonitrile-d3, 99.8 atom % d, acetonitrile-d isotopic 5g, cd3cn, methyl-d3 cyanide, trideuteroacetonitrile, 10mL, 02, IV, 428, 79.0 deg.C to 81.0 deg.C

For NMR
Color Colorless
Packaging Glass bottle
Water 0.1% max. (Coulometric)
Deuterium (D) 98.5% min.
Quantity 10mL
Viscosity 0.39 mPa.s (20°C)
Density 0.8440g/mL
Boiling Point 79.0°C to 81.0°C
Melting Point -46.0°C
Linear Formula CD3CN
Flash Point 2°C
Beilstein 02, IV, 428
Refractive Index 1.3430 to 1.3450
Formula Weight 44.07
Physical Form Liquid
Grade NMR
Chemical Name or Material Acetonitrile-d3, 99 atom%D
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger