Acetonitrile, Environmental Grade, 99.7+%, Alfa Aesar™

Suitable For Environmental, Pesticide Residue, and GC Analysis

Manufacturer: Alfa Aesar 40972K2

Quantity 1L

 View more versions of this product

Catalog No. AA40972K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-05-8
    Physical Form Liquid
    Assay Percent Range >99.7%
    Molecular Formula CH3CN
    Formula Weight 41.05
    MDL Number MFCD00001878
    Synonym Methyl cyanide
    Chemical Name or Material Acetonitrile
    Grade Environmental Grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Formula Weight 41.05
    Quantity 1L
    DOT Information DOT Class: 3, Packaging Group: II
    EINECS Number 200-835-2
    Ignition Point 525°C (977°F)
    Impurities Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05%, Halogenated residue 10ng/L (GC-ECD) 0.01%
    Odor Pungent
    Vapor Pressure 97 hPa (73mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range >99.7%
    Molecular Formula CH3CN
    MDL Number MFCD00001878
    CAS 75-05-8
    Synonym Methyl cyanide
    Health Hazard 1 H225-H302-H312-H332-H319: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.
    Health Hazard 2 Keep away from sources of ignition - No Smoking. Wear suitable protective clothing and gloves.
    Chemical Name or Material Acetonitrile
    Grade Environmental Grade

    Description

    Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05%, Halogenated residue 10ng/L (GC-ECD) 0.01%

    Keep away from sources of ignition - No Smoking. Wear suitable protective clothing and gloves.

    H225-H302-H312-H332-H319: Highly flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit