Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
CHEBI:38472
acetonitrile
CC#N

Function tested and optimized for LC/MS applications

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Formula Weight 41.05
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point 2°C (35.8°F)
Quantity 1L
Grade BAKER ANALYZED™ LC-MS Reagent
Chemical Name or Material Acetonitrile, Filtered through a 0.2 micron filter