Acetonitrile, GMP, High Purity Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 300000GMPPL05M

Packaging Metal Pail
Quantity 5 gal.

 View more versions of this product

Catalog No. 16-101-406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75-05-8
    Physical Form Liquid
    Molecular Formula C2H3N
    Formula Weight 41.05
    Synonym ACN
    Chemical Name or Material Acetonitrile

    Specifications

    Boiling Point 81° to 82°C
    Formula Weight 41.05
    Quantity 5 gal.
    Color Colorless, invisible vapor
    Packaging Metal Pail
    Flash Point 6°C (43°F)
    UN Number 1648; Colorless
    Physical Form Liquid
    CAS 75-05-8
    Synonym ACN
    Odor Ethereal
    Stability Stable under recommended storage conditions.
    Molecular Formula C2H3N
    Description General purpose organic solvent
    Chemical Name or Material Acetonitrile

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit