Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile (HPLC), Fisher Chemical™ Edge
Edge

Click to view available options
Quantity:
1L
2.5L
4L
19L
Packaging:
Amber Glass
NOWPak™ I
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N

HPLC mobile phase for analytical and preparative separations

Absorbance 0.01 max. at 254nm, 0.02 max. at 220nm, 0.04 max. at 210nm, 0.05 max. at 205nm, 0.07 max. at 200nm, 0.15 max. at 195nm, 1 max. at 190nm, Pass Test
Additional Information Vapor Pressure: 97mbar at 20°C
Assay Percent Range Assay: 99.9% min by GC
Linear Formula CH3CN
UN Number 1648
Merck Index 15, 69
Solubility Information Soluble in water
Formula Weight 41.05
Physical Form Liquid
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Description HPLC mobile phase for analytical and preparative separations. High UV transmittance and low evaporation residue. Suitable for gradient and isocratic conditions. Filtered through a 0.2Glc mum filter to remove particulates. This product also meets ACS specifications.
Fluorescence Background Pass Test
HPLC Gradient Suitability Pass Test
Identification Pass Test
LCMS Gradient Suitability Pass Test
Packaging Wide Neck Amber Bottle
Residue after Evaporation 1.0ppm max.
Specific Gravity 0.781
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
Water 0.01% max.
Flash Point 12.8°C (55°F)
Viscosity 0.36 cP at 20°C
Decomposition Information >500°C
Filtered Through 0.2-micron filtered.
Boiling Point 81°C
Color Colorless
Melting Point -46°C
Quantity 1L
Chemical Name or Material Acetonitrile
Show More Show Less

DANGER!

Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid