Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, LCMS, 99.9%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
75-05-8
75-05-8
CAS Max % 1
Aluminum (Al) 50 ppb
Barium (Ba) 5 ppb
Cadmium (Cd) 5 ppb
Calcium (Ca) 50 ppb
Percent Purity 0.999
Grade High Purity Solvent
Chromium (Cr) 5 ppb
Cobalt (Co) 5 ppb
Copper (Cu) 5 ppb
Iron (Fe) 5 ppb
Lead (Pb) 5 ppb
Magnesium (Mg) 10 ppb
Manganese (Mn) 5 ppb
Nickel (Ni) 5 ppb
Packaging Amber Glass Bottle
Potassium (K) 10 ppb
Refractive Index 1.3434-1.3446
Residue after Evaporation 1 ppm
Silver (Ag) 5 ppb
Sodium (Na) 50 ppb
Tin (Sn) 5 ppb
Water 0.0001
Zinc (Zn) 5 ppb
Quantity 1 L
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.