Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Reagent, ACS, 99.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
75-05-8
C2H3N
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
CC#N
75-05-8
C2H3N
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
CC#N

A1030, 75-05-8, CH3CN

Spectrum™ Acetonitrile, Reagent, ACS is a colorless liquid with low viscosity and low chemical reactivity and is used in chemical labs as a polar aprotic solvent in organic synthesis. Acetonitrile is a medium-polarity solvent with a convenient liquid range that is miscible with water. As an ACS grade Reagent, Spectrum Chemical manufactured acetonitrile is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
CAS Max % 1
Appearance Clear
UN Number UN1648
Percent Purity 99.5%
Packaging Drum
Residue after Evaporation 5e-005
Titratable Acid 8 μq/g
Titratable Base 0.6 μeq/g
Water 0.003
Quantity 200 L