Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Ultra Low Water, BakerDRY™, J.T.Baker™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N

Specifically manufactured for use in organometallic and oligonucleotide synthesis

Formula Weight 41.05
Grade BakerDRY™
Packaging Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point 2.00°C
Quantity 100mL
Chemical Name or Material Acetonitrile, Ultra Low Water